Kaliumdihydrogenphosphat 99+%, rein
Lieferant: Thermo Fisher Scientific
271080025
271080100
ACRO271080025EA
129
EUR
ACRO271080025
ACRO271080100
Kaliumdihydrogenphosphat 99+%, rein
Kaliumdihydrogenphosphat
Formel:
KH₂PO₄ Siedepunkt: ~ 100 °C (1013 hPa) Schmelzpunkt: 253 °C Dichte: 2,3 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011401 CAS-Nummer: 7778-77-0 EINECS: 231-913-4 REACH: 01-2119490224-41 Merck Index: 13,07744 |
Spezifikation Testergebnisse
Appearance | Colorless to white Crystalline powder or crystals |
Loss on drying | ≤1 % (105°C) |
Titration with NaOH | ≥99.0 % (on dried substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...