Sodium-2-propylvalerate 98%

Lieferant: Thermo Fisher Scientific
Gefahr

271830050 271830250
ACRO271830050EA 36.2 EUR
ACRO271830050 ACRO271830250
Sodium-2-propylvalerate 98%
Sodium-2-propylvalerate

Appearance: White Powder

Formel: C₈H₁₅O₂Na
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00078604
CAS-Nummer: 1069-66-5

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR