Boc-L-phenylalanin 99+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
N-Boc-L-phenylalanine, BOC-L-Phenylalanine, N-α-t-Boc-L-phenylalanine, Boc-Phe-OH
275640250
275640050
275641000
ACRO275640250EA
54.2
EUR
ACRO275640250
ACRO275640050
ACRO275641000
Boc-L-phenylalanin 99+%
Boc-L-phenylalanin
CAS No.: 13734-34-4
Formel:
C₁₄H₁₉NO₄ Schmelzpunkt: 85…88 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002663 CAS-Nummer: 13734-34-4 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 85 to 88 °C |
Water | ≤0.5 % (K.F.) |
HPLC | ≥99.0 % |
Specific optical rotation | +24° to +26° (20 °C, 589 nm) (c=1, ethanol) |
Titration with NaOH | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...