N-(tert-Butoxycarbonyl)-D-phenylalanine 99+%

Lieferant: Thermo Fisher Scientific

Synonyme: N-Boc-D-phenylalanine, BOC-D-Phenylalanine, Boc-D-Phe-OH

275740050 275740250
ACRO275740050EA 52.3 EUR
ACRO275740050 ACRO275740250
N-(tert-Butoxycarbonyl)-D-phenylalanine 99+%
N-(tert-Butoxycarbonyl)-D-phenylalanine

CAS No.: 18942-49-9

Formel: C₁₄H₁₉NO₄
Schmelzpunkt: 78…87 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00063149
CAS-Nummer: 18942-49-9

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 85 to 88 °C
Water ≤0.5 % (K.F.)
HPLC ≥99.0 %
Specific optical rotation -24° to -26° (20 °C, 589 nm) (c=1, ethanol)
Titration with NaOH ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR