Wasser, pure_f, demineralized
Lieferant: Thermo Fisher Scientific
Synonyme:
Wasserstoffoxid, Hydrogenoxid
276010050
276010200
ACRO276010050EA
64.6
EUR
ACRO276010050
ACRO276010200
Wasser, pure_f, demineralized
Wasser
Formel:
H₂O Molecular Weight: 18.02 g/mol Siedepunkt: 100 Schmelzpunkt: 0 Dichte: 1 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011332 CAS-Nummer: 7732-18-5 EINECS: 231-791-2 Merck Index: 13,10098 |
Spezifikation Testergebnisse
Appearance | Clear colorless Liquid |
pH | 6 to 8 |
Silicon dioxide (SiO2) | ≤0.1 ppm |
Conductivity | ≤5.0 µS/cm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...