Polyvinylpyrrolidon, M.W. ∼ 3500 (K12)

Lieferant: Thermo Fisher Scientific

276142500 276140010
ACRO276142500EA 54 EUR
ACRO276142500 ACRO276140010
Polyvinylpyrrolidon, M.W. ∼ 3500 (K12)
Polyvinylpyrrolidon
Formel: (C₆H₉NO)n
Lagertemperatur: Raumtemperatur
MDL: MFCD01076626
CAS-Nummer: 9003-39-8

Bestellen

Spezifikation Testergebnisse

Appearance White to yellowish-white Powder or flakes
Infrared spectrum Conforms
Water ≤5.0 % (K.F.)
Sulfated ash ≤0.1 %
Additional info European color test - R color: R6
HPLC ≤3.0 % 2-Pyrrolidone
Identification Passes test
pH 3.0 to 5.0 (5 % in water)
Nitrogen (N) 11.5 to 12.8 % (on anhydrous substance)
Heavy metals (as Pb) ≤10 ppm
Aldehyde ≤0.05 %
Hydrazine (N2H4) ≤1 ppm
Peroxides ≤400 ppm (Titanyl sulphate method)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR