9-Methylacridin 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
276490010
ACRO276490010EA
162
EUR
ACRO276490010
9-Methylacridin 97%
9-Methylacridine
Formel:
C₁₄H₁₁N Schmelzpunkt: 114…119 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143523 CAS-Nummer: 611-64-3 EINECS: 210-272-4 |
Spezifikation Testergebnisse
Appearance | Yellow to light brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 114 to 119 °C |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...