1-Methyl-2-benzimidazolinone 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1,3-Dihydro-1-methyl-2H-benzimidazol-2-one, 1-Methyl-1H-benzo[d]imidazol-2(3H)-one

276500050
ACRO276500050EA 148 EUR
ACRO276500050
1-Methyl-2-benzimidazolinone 98%
1-Methyl-2-benzimidazolinone

Appearance: White to light beige Crystalline powder

Formel: C₈H₈N₂O
Molecular Weight: 148,165 g/mol
Schmelzpunkt: 192…196 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00142654
CAS-Nummer: 1849-01-0

Bestellen

Spezifikation Testergebnisse

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 192 to 196 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR