1-Methyl-2-formylbenzimidazole 99+%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-Methyl-1H-benzo[d]imidazole-2-carbaldehyde

276520010
ACRO276520010EA 94.6 EUR
ACRO276520010
1-Methyl-2-formylbenzimidazole 99+%
1-Methyl-2-formylbenzimidazole

Appearance: Off-white to light beige Crystalline powder

Formel: C₉H₈N₂O
Molecular Weight: 160,176 g/mol
Schmelzpunkt: 114…116 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00142655
CAS-Nummer: 3012-80-4

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 119 to 122 °C
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR