(R)-(+)-1-Amino-2-(methoxymethyl)pyrrolidine 95+%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
RAMP
277140010
ACRO277140010EA
130
EUR
ACRO277140010
(R)-(+)-1-Amino-2-(methoxymethyl)pyrrolidine 95+%
(R)-(+)-1-Amino-2-(methoxymethyl)pyrrolidine
Appearance: Clear colorless Liquid
Formel:
C₆H₁₄N₂O Molecular Weight: 130,19 g/mol Siedepunkt: 42 °C (1,8 mmHg) Dichte: 0,97 g/cm³ Flammpunkt: 72 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00010622 CAS-Nummer: 72748-99-3 |
Spezifikation Testergebnisse
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4635 to 1.4655 (20°C, 589 nm) |
GC | ≥95.0 % |
Specific optical rotation | +76° to +84° (20°C, 589 nm) (neat) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...