Titan(IV)oxid 98,0-100,5%
Lieferant: Thermo Fisher Scientific
277370010
277370100
ACRO277370010EA
66.6
EUR
ACRO277370010
ACRO277370100
Titan(IV)oxid 98,0-100,5%
Titan(IV)oxid
Formel:
TiO₂ Siedepunkt: 2900 °C (760 mmHg) Schmelzpunkt: 1855 °C Dichte: 4,23 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011269 CAS-Nummer: 13463-67-7 EINECS: 236-675-5 Merck Index: 13,09549 |
Spezifikation Testergebnisse
Appearance | White to almost white Powder |
Acidity/alkalinity | Passes test |
Titration | 98.0 to 100.5 % |
Antimony (Sb) | ≤100 ppm |
Arsenic (As) | ≤5 ppm |
Barium (Ba) | passes test |
Iron (Fe) | ≤200 ppm |
Soluble matter | ≤0.5 % (in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...