7-Hydroxy-6-methoxy-3,4-dihydroisoquinoline 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
277822500
ACRO277822500EA
49.5
EUR
ACRO277822500
7-Hydroxy-6-methoxy-3,4-dihydroisoquinoline 98%
7-Hydroxy-6-methoxy-3,4-dihydroisoquinoline
Appearance: Yellow Powder and/or flakes
Formel:
C₁₀H₁₁NO₂ Schmelzpunkt: 187…191 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143517 CAS-Nummer: 4602-73-7 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | Yellow Powder and/or flakes |
Infrared spectrum | Conforms |
Melting point | 187 to 191 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...