Polysorbat 60
Lieferant: Thermo Fisher Scientific
278620010
278622500
278620025
ACRO278620010EA
80.3
EUR
ACRO278620010
ACRO278622500
ACRO278620025
Polysorbat 60
Polysorbat 60
Formel:
C₆₄H₁₂₆O₂₆ Siedepunkt: ? 110 °C (1013 hPa) Dichte: 1,08 g/cm³ Flammpunkt: >150 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01780110 CAS-Nummer: 9005-67-8 EINECS: 500-020-4 |
Spezifikation Testergebnisse
Appearance | Light yellow to light brown Paste |
Infrared spectrum | Conforms |
Water | ≤3 % |
Dioxane | ≤5 ppm |
Acid value | ≤2 mg KOH/g |
Hydroxyl value | 81 to 96 mg KOH/g |
Saponification value | 45 to 55 mg KOH/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...