2-Chloro-2-ethoxyacetic acid ethyl ester 90% remainder mainly ethyl diethoxyacetate

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Ethyl 2-chloro-2-ethoxyacetate, Ethyl chloro(ethoxy)acetate

279870500
ACRO279870500EA 151 EUR
ACRO279870500
2-Chloro-2-ethoxyacetic acid ethyl ester 90% remainder mainly ethyl diethoxyacetate
2-Chloro-2-ethoxyacetic acid ethyl ester

Appearance: Clear colorless to orange-brown Liquid

Formel: C₆H₁₁ClO₃
Molecular Weight: 166.601 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00043953
CAS-Nummer: 34006-60-5

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to orange-brown Liquid
Infrared spectrum Conforms
GC ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR