(+)-2-Methyl-L-serine 99%
Lieferant: Thermo Scientific
Synonyme:
(S)-2-Amino-3-hydroxy-2-methylpropanoic acid, H-α-Me-Ser-OH
290012500
ACRO290012500EA
156
EUR
ACRO290012500
(+)-2-Methyl-L-serine 99%
(+)-2-Methyl-L-serine
Appearance: White Powder
Formel:
C₄H₉NO₃ Molecular Weight: 119,12 g/mol Schmelzpunkt: 286…288 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00145243 CAS-Nummer: 16820-18-1 |
Spezifikation Testergebnisse
Appearance | White Powder |
Infrared spectrum | Conforms |
TLC | ≥98.5 % |
Specific optical rotation | +2.3° to +4.3° (25°C, 589 nm) (c=5, 6 N HCl) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...