3-phenylpyrrolidine-2-carboxylic acid 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(2S,3R)-3-Phenylpyrrolidine-2-carboxylic acid
290081000
ACRO290081000EA
179
EUR
ACRO290081000
3-phenylpyrrolidine-2-carboxylic acid 98%
3-phenylpyrrolidine-2-carboxylic acid
Appearance: White to off-white Powder
Formel:
C₁₁H₁₃NO₂ Schmelzpunkt: 234…236 °C Lagertemperatur: Raumtemperatur |
CAS-Nummer:
118758-48-8 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Specific optical rotation | +82° to +86° (25°C, 589 nm) (c=0.75, 6 N HCl) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...