4-Chlor-2-fluorbenzaldehyd 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
290620050
290620250
ACRO290620050EA
96
EUR
ACRO290620050
ACRO290620250
4-Chlor-2-fluorbenzaldehyd 99%
4-Chloro-2-fluorobenzaldehyde
Formel:
C₇H₄ClFO Molecular Weight: 158.56 g/mol Schmelzpunkt: 58…61 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143282 CAS-Nummer: 61072-56-8 |
Spezifikation Testergebnisse
Appearance | White to yellow to light beige Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 57 to 61 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...