Octafluornaphthalin 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Perfluoronaphthalene
291530010
ACRO291530010EA
52.7
EUR
ACRO291530010
Octafluornaphthalin 96%
Octafluornaphthalin
Formel:
C₁₀F₈ Molecular Weight: 272.1 g/mol Siedepunkt: 238 Schmelzpunkt: 85…90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014307 CAS-Nummer: 313-72-4 EINECS: 206-239-9 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 85 to 90 °C |
GC | ≥95 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...