[2.2]-Paracyclophan 99%

Lieferant: Thermo Fisher Scientific

291600010 291600250 291600050
ACRO291600010EA 24.1 EUR
ACRO291600010 ACRO291600250 ACRO291600050
[2.2]-Paracyclophan 99%
[2.2]-Paracyclophan
Formel: C₁₆H₁₆
Schmelzpunkt: 285…288 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003707
CAS-Nummer: 1633-22-3

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 285 to 288 °C
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR