Kaliumsorbat 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
291740010
291742500
291740050
291741000
ACRO291740010EA
97.4
EUR
ACRO291740010
ACRO291742500
ACRO291740050
ACRO291741000
Kaliumsorbat 99%
Kaliumsorbat
Formel:
CH₃CH=CHCH=CHCOOK Schmelzpunkt: 270 °C (Zersetzung) Dichte: 1,36 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016546 CAS-Nummer: 24634-61-5 EINECS: 246-376-1 Merck Index: 13,07756 |
|
Spezifikation Testergebnisse
Appearance | White to cream Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤1 % (105°C, 3 h) |
Acidity/alkalinity | passes test |
Titration with HClO4 | 98.5 to 101.5 % |
Arsenic (As) | ≤3 ppm |
Lead (Pb) | ≤2 ppm |
Mercury (Hg) | ≤1 ppm |
Heavy metals (as Pb) | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...