2-Chlorbenzimidazol 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Chlor-1H-benzimidazol
291960250
AC29196.1
AC29196.10
AC29196.50
ACRO291960250EA
78.4
EUR
ACRO291960250
MAYBAC29196.1
MAYBAC29196.10
MAYBAC29196.50
2-Chlorbenzimidazol 97%
2-Chlorobenzimidazole
Formel:
C₇H₅ClN₂ Molecular Weight: 152.58 g/mol Schmelzpunkt: 200 °C |
MDL:
MFCD00051944 CAS-Nummer: 4857-06-1 EINECS: 225-453-3 |
Spezifikation Testergebnisse
Appearance | Beige to brown Crystalline powder |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...