5,10,15,20-Tetrakis(pentafluorophenyl)porphyrin 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5,10,15,20-Tetrakis(perfluorophenyl)porphyrin, 5,10,15,20-Tetrakis(pentafluorophenyl)-21H,23H-porphine, 5,10,15,20-Tetrakis(pentafluorophenyl)-21h,23h-porphine

292151000
ACRO292151000EA 118 EUR
ACRO292151000
5,10,15,20-Tetrakis(pentafluorophenyl)porphyrin 95%
5,10,15,20-Tetrakis(pentafluorophenyl)porphyrin

Appearance: Violet to purple Crystalline powder

Formel: C₄₄H₁₀F₂₀N₄
Molecular Weight: 974,56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00010032
CAS-Nummer: 25440-14-6

Bestellen

Spezifikation Testergebnisse

Appearance Violet to purple Crystalline powder
Infrared spectrum Conforms
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR