2,5-Pyrazinedicarboxylic acid dihydrate 97%

Lieferant: Thermo Scientific
Achtung

292320010
ACRO292320010EA 113 EUR
ACRO292320010
2,5-Pyrazinedicarboxylic acid dihydrate 97%
2,5-Pyrazinedicarboxylic acid dihydrate

Appearance: White to light beige Powder

Formel: C₆H₆N₂O₅
Molecular Weight: 186,123 g/mol
Schmelzpunkt: 257 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00216919
CAS-Nummer: 205692-63-3

Bestellen

Spezifikation Testergebnisse

Appearance White to light beige Powder
Infrared spectrum Conforms
Water 16 to 19 % (K.F.)
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR