2-Nitrophenyl selenocyanate 98%

Lieferant: Thermo Fisher Scientific
Gefahr

293580010
ACRO293580010EA 56 EUR
ACRO293580010
2-Nitrophenyl selenocyanate 98%
2-Nitrophenyl selenocyanate

Appearance: Brown Crystalline powder

Formel: C₇H₄N₂O₂Se
Schmelzpunkt: 138…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00043146
CAS-Nummer: 51694-22-5
UN: 3283
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to brown Powder or crystals or chunks
Infrared spectrum Conforms
Melting point 139 to 145 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR