(R)-(-)-3-Chinuclidinol 99+%
Lieferant: Thermo Scientific
|
Gefahr
|
293730025
ACRO293730025EA
98.8
EUR
ACRO293730025
(R)-(-)-3-Chinuclidinol 99+%
(R)-(-)-3-Chinuclidinol
Formel:
C₇H₁₃NO Molecular Weight: 127,19 g/mol Siedepunkt: 120 °C (27mbar) Schmelzpunkt: 217…224 °C |
CAS-Nummer:
25333-42-0 EINECS: 246-857-6 UN: 3263 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | White to light beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 217 to 224 °C |
GC | ≥99.0 % |
Specific optical rotation | -43° to -46° (20 °C, 589 nm) (c=3, 1 N HCl) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...