4-Methoxy-2,6-bis(2-methyl-2-propanyl)phenol 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3,5-Di-tert-butyl-4-hydroxyanisol
294160250
294161000
ACRO294160250EA
63.3
EUR
ACRO294160250
ACRO294161000
4-Methoxy-2,6-bis(2-methyl-2-propanyl)phenol 97%
2,6-Di-tert-butyl-4-methoxyphenol
Formel:
C₁₅H₂₄O₂ Molecular Weight: 236.35 g/mol Siedepunkt: 135…140 °C (10 mmHg) Schmelzpunkt: 103…106 °C |
MDL:
MFCD00008824 CAS-Nummer: 489-01-0 EINECS: 207-693-0 |
Spezifikation Testergebnisse
Appearance | White to beige Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 102 to 106 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...