3-Cyclohexen-1-carbonsäure 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Cyclohex-3-enecarboxylic acid, 3-Cyclohexenecarboxylic acid, 1,2,3,6-Tetrahydrobenzoic Acid

294390100 294390010
ACRO294390100EA 176 EUR
ACRO294390100 ACRO294390010
3-Cyclohexen-1-carbonsäure 97%
3-Cyclohexen-1-carbonsäure

Appearance: Clear yellow-brown to blue-grey Liquid

Formel: C₇H₁₀O₂
Molecular Weight: 126.16 g/mol
Siedepunkt: 238 °C (760 mmHg)
Schmelzpunkt: 17 °C
Dichte: 1.08 g/cm³
Flammpunkt: >112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013781
CAS-Nummer: 4771-80-6
UN: 3265
ADR: 8 II

Bestellen

Spezifikation Testergebnisse

Appearance Clear yellow-brown to blue-grey Liquid
Infrared spectrum Conforms
Refractive index 1.4800 to 1.4820 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR