(1R,2R)-(+)-N,N'-Dimethyl-1,2-diphenyl-1,2-ethane diamine 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
294662500
ACRO294662500EA
143
EUR
ACRO294662500
(1R,2R)-(+)-N,N'-Dimethyl-1,2-diphenyl-1,2-ethane diamine 99%
(1R,2R)-(+)-N,N'-Dimethyl-1,2-diphenyl-1,2-ethane diamine
Appearance: White Crystals
Formel:
C₁₆H₂₀N₂ Schmelzpunkt: 48…51 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00216955 CAS-Nummer: 118628-68-5 UN: 3259 ADR: 8,III |
Spezifikation Testergebnisse
Appearance | White to yellow Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 48 to 51 °C |
Specific optical rotation | +48° to +51° (20 °C, 589 nm) (c=2, MeOH) |
Titration with HClO4 | 98.5 to 101.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...