Bortribromid 99.9%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
295200250
295201000
295205000
ACRO295200250EA
78.5
EUR
ACRO295200250
ACRO295201000
ACRO295205000
Bortribromid 99.9%
Bortribromid
Formel:
BBr₃ Siedepunkt: 91,2 °C (760 mmHg) Schmelzpunkt: –46 °C Dichte: 2,65 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011312 CAS-Nummer: 10294-33-4 EINECS: 233-657-9 UN: 2692 ADR: 8,I Merck Index: 13,01337 |
Spezifikation Testergebnisse
Appearance | Clear amber Liquid |
Assay | ≥99.85 % (By difference) |
Titration | ≥99.2 % |
Sulfur (S) | ≤100 ppm |
Heavy metals (as Pb) | ≤100 ppm |
Impurity | ≤0.13 % Free bromine (Br2) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...