2,6-Diaminopurin 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
9H-Purine-2,6-diamine, 7H-Purine-2,6-diamine
295270050
ACRO295270050EA
74.4
EUR
ACRO295270050
2,6-Diaminopurin 98%
2,6-Diaminopurine
Formel:
C₅H₆N₆ Molecular Weight: 150,14 g/mol Schmelzpunkt: 302 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00213668 CAS-Nummer: 1904-98-9 EINECS: 217-605-2 |
Spezifikation Testergebnisse
Appearance | White to light yellow Crystalline powder or granules |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...