3-Phenyl-1H-pyrazole-4-carbaldehyde 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Phenyl-1H-pyrazole-4-carboxaldehyde
296000010
ACRO296000010EA
108
EUR
ACRO296000010
3-Phenyl-1H-pyrazole-4-carbaldehyde 98%
3-Phenyl-1H-pyrazole-4-carbaldehyde
Appearance: Light yellow Powder
Formel:
C₁₀H₈N₂O Molecular Weight: 172,187 g/mol Schmelzpunkt: 146…148 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00216971 CAS-Nummer: 26033-20-5 |
Spezifikation Testergebnisse
Appearance | Light yellow Powder |
Infrared spectrum | Conforms |
Melting point | 141 to 146 °C |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...