Potassium tris(1-pyrazolyl)borohydride 93%

Lieferant: Thermo Scientific
Achtung

Synonyme: Potassium Hydrotris(1-pyrazolyl)borate

296010050
ACRO296010050EA 127 EUR
ACRO296010050
Potassium tris(1-pyrazolyl)borohydride 93%
Potassium tris(1-pyrazolyl)borohydride

Appearance: White Crystalline powder

Formel: C₉H₁₀BKN₆
Molecular Weight: 252,13 g/mol
Schmelzpunkt: 182…188 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00040346
CAS-Nummer: 18583-60-3
UN: 2813
ADR: 4.3,III

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 182 to 188 °C
Titration with HClO4 ≥91 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR