Potassium tris(3,5-dimethylpyrazol-1-yl)borohydride 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Potassium Hydrotris(3,5-dimethylpyrazol-1-yl)borate, Potassium hydrotris(3,5-dimethylpyrazol-1-yl)borate
296020050
ACRO296020050EA
127
EUR
ACRO296020050
Potassium tris(3,5-dimethylpyrazol-1-yl)borohydride 99%
Potassium tris(3,5-dimethylpyrazol-1-yl)borohydride
Appearance: White Powder or needles
Formel:
C₁₅H₂₂BKN₆ Molecular Weight: 336,29 g/mol Schmelzpunkt: 287 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040342 CAS-Nummer: 17567-17-8 UN: 2813 ADR: 4.3,III |
Spezifikation Testergebnisse
Appearance | White Powder or needles |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...