Methansulfonamid 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Methylsulphonamide
296110250
296111000
ACRO296110250EA
55.8
EUR
ACRO296110250
ACRO296111000
Methansulfonamid 98%
Methansulfonamid
Formel:
CH₅NO₂S Schmelzpunkt: 88…92 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007940 CAS-Nummer: 3144-09-0 EINECS: 221-553-6 |
Spezifikation Testergebnisse
Appearance | White to yellow Powder or crystals or flakes |
Infrared spectrum | Conforms |
Melting point | 88 to 92 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...