Methansulfonamid 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Methylsulphonamide

296110250 296111000
ACRO296110250EA 55.8 EUR
ACRO296110250 ACRO296111000
Methansulfonamid 98%
Methansulfonamid
Formel: CH₅NO₂S
Schmelzpunkt: 88…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007940
CAS-Nummer: 3144-09-0
EINECS: 221-553-6

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Powder or crystals or flakes
Infrared spectrum Conforms
Melting point 88 to 92 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR