2-(1-benzylpiperidin-4-yl)ethanol 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: N-Benzyl-4-(2-hydroxyethyl)piperidine

296480010
ACRO296480010EA 65.8 EUR
ACRO296480010
2-(1-benzylpiperidin-4-yl)ethanol 97%
2-(1-benzylpiperidin-4-yl)ethanol

Appearance: Yellow to light brown Viscous liquid

Formel: C₁₄H₂₁NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00800492
CAS-Nummer: 76876-70-5

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to light brown Viscous liquid
Infrared spectrum Conforms
Refractive index 1.5370 to 1.5400 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR