Pyrazol-3,5-dicarbonsäure-diethylester 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: Pyrazole-3,5-dicarboxylic Acid Diethyl Ester, Diethyl 3,5-pyrazoledicarboxylate

296520050
ACRO296520050EA 87.4 EUR
ACRO296520050
Pyrazol-3,5-dicarbonsäure-diethylester 99%
Diethyl pyrazole-3,5-dicarboxylate
Formel: C₉H₁₂N₂O₄
Molecular Weight: 212,21 g/mol
Schmelzpunkt: 51…55 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00152167
CAS-Nummer: 37687-24-4

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 51 to 58 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR