4-(4-Chlorphenyl)-4-piperidinol 99+%, kristallin

Lieferant: Thermo Fisher Scientific
Achtung

296840010 296840050 296840250
ACRO296840010EA 26.1 EUR
ACRO296840010 ACRO296840050 ACRO296840250
4-(4-Chlorphenyl)-4-piperidinol 99+%, kristallin
4-(4-Chlorphenyl)-4-piperidinol
Formel: C₁₁H₁₄ClNO
Schmelzpunkt: 139…141 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006001
CAS-Nummer: 39512-49-7
EINECS: 254-479-8

Bestellen

Spezifikation Testergebnisse

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point 137 to 141 °C
Loss on drying ≤0.5 % (1 g, 105 °C, 2 h, 760 mmHg)
Sulfated ash ≤0.2 %
TLC ≤0.5 % (Each individual impurity)
UV at 440 nm A: ≤0.05
Titration with HClO4 98.0 to 102.0 % (On dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR