Glycin-tert-butylester Hydrochlorid (H-Gly-OtBu.HCl) 99%

Lieferant: Thermo Scientific

Synonyme: H-Gly-OtBu.HCl, Glycine tert-butyl ester hydrochloride, H-Gly-OtBu·HCl

297020025
ACRO297020025EA 91 EUR
ACRO297020025
Glycin-tert-butylester Hydrochlorid (H-Gly-OtBu.HCl) 99%
Glycin-tert-butylester Hydrochlorid (H-Gly-OtBu.HCl)
Formel: C₆H₁₄ClNO₂
Molecular Weight: 167,64 g/mol
Schmelzpunkt: 138…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00058255
CAS-Nummer: 27532-96-3

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
TLC ≥98.5 %
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR