trans-2-Decenal 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(E)-2-Decenal
298190250
ACRO298190250EA
92.7
EUR
ACRO298190250
trans-2-Decenal 95%
trans-2-Decenal
Formel:
C₁₀H₁₈O Molecular Weight: 154,25 g/mol Siedepunkt: 78…80 °C (3 mmHg) Flammpunkt: 96 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00014679 CAS-Nummer: 3913-81-3 EINECS: 223-474-2 UN: 3082 ADR: 9,III |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4510 to 1.4600 (20°C, 589 nm) |
GC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...