cis-2-Amino-1-cyclopentanecarboxylic acidhydrochloride hemihydrate 99%

Lieferant: Thermo Scientific
Gefahr

298382500
ACRO298382500EA 60.4 EUR
ACRO298382500
cis-2-Amino-1-cyclopentanecarboxylic acidhydrochloride hemihydrate 99%
cis-2-Amino-1-cyclopentanecarboxylic acidhydrochloride hemihydrate

Appearance: White to off-white Crystalline powder

Formel: C₆H₁₄ClNO₃
Schmelzpunkt: 199…202 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00800502
CAS-Nummer: 212755-84-5
UN: 3261
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤6 % (K.F.)
Titration with HClO4 ≥98.5 % (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR