Ethyl-cis-2-amino-1-cyclopentane carboxylate hydrochloride 99%

Lieferant: Thermo Scientific

Synonyme: (1R,2S)-rel-Ethyl 2-aminocyclopentanecarboxylate hydrochloride

298392500
ACRO298392500EA 36.6 EUR
ACRO298392500
Ethyl-cis-2-amino-1-cyclopentane carboxylate hydrochloride 99%
Ethyl-cis-2-amino-1-cyclopentane carboxylate hydrochloride

Appearance: White Crystalline powder

Formel: C₈H₁₆ClNO₂
Molecular Weight: 193,671 g/mol
Schmelzpunkt: 97…100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00800503
CAS-Nummer: 142547-15-7

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 97 to 100 °C
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR