trans-2-(4-Bromobenzoyl)-1-cyclohexanecarboxylic acid 98%

Lieferant: Thermo Scientific

298690010
ACRO298690010EA 202 EUR
ACRO298690010
trans-2-(4-Bromobenzoyl)-1-cyclohexanecarboxylic acid 98%
trans-2-(4-Bromobenzoyl)-1-cyclohexanecarboxylic acid

Appearance: White Crystalline powder

Formel: C₁₄H₁₅BrO₃
Schmelzpunkt: 163…169 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00800528
CAS-Nummer: 212757-09-0

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 163 to 169 °C
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR