Bis(triphenylphosphin)palladium(II)chlorid 98%

Lieferant: Thermo Fisher Scientific
Achtung

299250025 299250050 299250250 299251000
ACRO299250025EA 124 EUR
ACRO299250025 ACRO299250050 ACRO299250250 ACRO299251000
Bis(triphenylphosphin)palladium(II)chlorid 98%
Bis(triphenylphosphin)palladium(II)chlorid
Formel: C₃₆H₃₀Cl₂P₂Pd
Schmelzpunkt: 260 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009593
CAS-Nummer: 13965-03-2
EINECS: 237-744-2

Bestellen

Spezifikation Testergebnisse

Appearance Yellow Powder or crystals and/or chunks
Infrared spectrum Conforms
Assay 14.8 to 15.5 % (Pd)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR