cis-2-(tert-Butoxycarbonylamino)-1-cyclopentanecarboxylic acid 98%

Lieferant: Thermo Scientific

Synonyme: (1R,2S)-2-(Boc-amino)cyclopentanecarboxylic Acid, cis-2-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid

299582500
ACRO299582500EA 74.5 EUR
ACRO299582500
cis-2-(tert-Butoxycarbonylamino)-1-cyclopentanecarboxylic acid 98%
cis-2-(tert-Butoxycarbonylamino)-1-cyclopentanecarboxylic acid

CAS No.: 136315-70-3

Formel: C₁₁H₁₉NO₄
Molecular Weight: 229,276 g/mol
Schmelzpunkt: 80…86 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00800539
CAS-Nummer: 136315-70-3

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR