L-Prolinamid 98%
Lieferant: Thermo Fisher Scientific
302990010
302990050
ACRO302990010EA
43.7
EUR
ACRO302990010
ACRO302990050
L-Prolinamid 98%
L-Prolinamide
CAS No.: 7531-52-4
Formel:
C₅H₁₀N₂O Schmelzpunkt: 97…102 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00005253 CAS-Nummer: 7531-52-4 |
Spezifikation Testergebnisse
Appearance | White to light yellow Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 96 to 104 °C |
HPLC | ≥97.5 % |
Specific optical rotation | -103° to -111° (20 °C, 589 nm) (c=1, EtOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...