N,N,N′,N′-Tetramethyl-O-(N-succinimidyl)-uroniumtetrafluoroborat 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate, TSTU, 2-(2,5-Dioxopyrrolidin-1-yl)-1,1,3,3-tetramethylisouronium tetrafluoroborate
303190010
303190050
ACRO303190010EA
38.1
EUR
ACRO303190010
ACRO303190050
N,N,N′,N′-Tetramethyl-O-(N-succinimidyl)-uroniumtetrafluoroborat 97%
N,N,N′,N′-Tetramethyl-O-(N-succinimidyl)-uroniumtetrafluoroborat
Formel:
C₉H₁₆BF₄N₃O₃ Molecular Weight: 301.05 g/mol Schmelzpunkt: 194…200 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00077875 CAS-Nummer: 105832-38-0 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder or needles |
Infrared spectrum | Conforms |
Elemental Analysis | 13.40 to 14.52 % (N) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...