3-Fluor-4-nitrophenol 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
303370050
303370010
ACRO303370050EA
102
EUR
ACRO303370050
ACRO303370010
3-Fluor-4-nitrophenol 99%
3-Fluor-4-nitrophenol
Formel:
C₆H₄FNO₃ Schmelzpunkt: 92…95 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041251 CAS-Nummer: 394-41-2 EINECS: 206-895-6 UN: 2811 |
Spezifikation Testergebnisse
Appearance | Light yellow to brown Powder |
Infrared spectrum | Conforms |
Melting point | 91 to 95 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...