4-tert-Butylbenzolsulfonylchlorid 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
303470100
303470500
ACRO303470100EA
48.9
EUR
ACRO303470100
ACRO303470500
4-tert-Butylbenzolsulfonylchlorid 98%
4-tert-Butylbenzolsulfonylchlorid
Formel:
C₁₀H₁₃ClO₂S Siedepunkt: 165 °C (18 mmHg) Schmelzpunkt: 79…83 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007449 CAS-Nummer: 15084-51-2 UN: 3261 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | White to off-white to light yellow Powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 78 to 84 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...