Sodium metatungstate hydrate, kristallin

Lieferant: Thermo Fisher Scientific
Gefahr

304090500 304092500 304090010
ACRO304090500EA 82.7 EUR
ACRO304090500 ACRO304092500 ACRO304090010
Sodium metatungstate hydrate, kristallin
Sodium metatungstate hydrate

CAS No.: 652154-12-6

Formel: H₂Na₆O₄₀W₁₂·xH₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00150663
CAS-Nummer: 652154-12-6

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder or beads
Assay ≥85 % (WO3)
Stabilizer ≤200 ppm (Cr-salt) Typical


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR